(R)-(-)-1-Aminopropan-2-ol 95%
Supplier: Ambeed
|
Danger
|
A250858-1G
A250858-25G
A250858-5G
A250858-250MG
103811-954EA
12
USD
103811-954
103851-794
77091-162
77091-164
77091-166
77098-684
(R)-(-)-1-Aminopropan-2-ol 95%
(R)-(-)-1-Aminopropan-2-ol
Precaución: for Research Use Only
Formula:
C₃H₉NO Boiling Pt: 159.9 °C (760 mm Hg) Melting Pt: 24…26 °C Flash Pt: 71 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00064428 CAS Number: 2799-16-8 UN: 2735 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...