(R)-1,4-Diazabicyclo[4.3.0]nonane 95%

Supplier: Ambeed
Danger

A178297-10G A178297-250MG A178297-1G A178297-5G A178297-100MG
103807-316EA 43 USD
103807-316 103845-656 77029-330 77029-332 77083-068 77083-070 77871-892
(R)-1,4-Diazabicyclo[4.3.0]nonane 95%
(R)-1,4-Diazabicyclo[4.3.0]nonane

Precaución: for Research Use Only

Formula: C₇H₁₄N₂
Boiling Pt: 199.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09752820
CAS Number: 96193-27-0
UN: 3267
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR