(R)-(-)-1,2,3,4-Tetrahydro-1-naphthylamine 98%

Supplier: Ambeed
Danger

Synonyms: (R)-1,2,3,4-Tetrahydronaphthalen-1-amine

A236877-1G A236877-5G A236877-100G A236877-10G A236877-25G
103811-120EA 5 USD
103811-120 103850-892 77089-952 77089-954 77140-174 77140-176 77304-064
(R)-(-)-1,2,3,4-Tetrahydro-1-naphthylamine 98%
(R)-(-)-1,2,3,4-Tetrahydro-1-naphthylamine

Precaución: for Research Use Only

Formula: C₁₀H₁₃N
MW: 147.22 g/mol
Boiling Pt: 118…120 °C (8 mm Hg)
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00671629
CAS Number: 23357-46-2
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR