(R)-1-Boc-4-Oxopiperidine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: (2R)-4-Oxopiperidine-2-carboxylic acid, N-BOC protected, (R)-1-(tert-Butoxycarbonyl)-4-oxopiperidine-2-carboxylic acid

A610277-5G A610277-10G A610277-250MG
103840-162EA 167 USD
103840-162 76844-328 77018-040 77018-042
(R)-1-Boc-4-Oxopiperidine-2-carboxylic acid 95%
(R)-1-Boc-4-Oxopiperidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₅
MW: 243.26 g/mol
Melting Pt: 119…124 °C
Storage Temperature: Ambient
MDL Number: MFCD09953681
CAS Number: 1212176-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR