(R)-1-(pyridin-4-yl)ethanol 98%

Supplier: Ambeed
Warning

Synonyms: (R)-alpha-Methyl-4-pyridinemethanol, (R)-1-(4-Pyridyl)ethanol, (R)-4-(1-Hydroxyethyl)pyridine, (R)-1-(Pyridin-4-yl)ethanol

A244021-1G A244021-5G A244021-100MG A244021-250MG A244021-25G
103811-562EA 17 USD
103811-562 103851-386 77090-590 77090-592 77140-334 77140-336 77140-338
(R)-1-(pyridin-4-yl)ethanol 98%
(R)-1-(pyridin-4-yl)ethanol

Precaución: for Research Use Only

Formula: C₇H₉NO
MW: 123.155 g/mol
Boiling Pt: 239.7 °C (760 mm Hg)
Melting Pt: 67…69 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077865
CAS Number: 27854-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR