(R)-2-(1-Hydroxyethyl)pyridine 95%

Supplier: Ambeed
Warning

Synonyms: (R)-1-(Pyridin-2-yl)ethanol

A221447-5G A221447-250MG A221447-25G
103818-924EA 33 USD
103818-924 103849-804 77088-402 77097-968 77097-970
(R)-2-(1-Hydroxyethyl)pyridine 95%
(R)-2-(1-Hydroxyethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₉NO
MW: 123.155 g/mol
Boiling Pt: 210.6 °C (760 mm Hg)
Flash Pt: 81.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04972322
CAS Number: 27911-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR