(R)-1-(Pyridin-2-yl)ethanamine 98%

Supplier: Ambeed
Warning

Synonyms: (+)-2-[(1R)-1-Aminoethyl]pyridine

A148631-250MG A148631-1G A148631-5G
103803-952EA 47 USD
103803-952 103842-068 77137-678 77317-528 77317-530
(R)-1-(Pyridin-2-yl)ethanamine 98%
(R)-1-(Pyridin-2-yl)ethanamine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MW: 122.1677 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07776289
CAS Number: 45695-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR