(R)-(+)-1-(1-Naphthyl)ethylamine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-(+)-α-(1-Naphthyl)ethylamine, (R)-1-(Naphthalen-1-yl)ethanamine, (+)-1-(1-Naphthyl)ethylamine
A311072-25G
A311072-100G
A311072-10G
A311072-1G
A311072-5G
A311072-500G
103835-838EA
7
USD
103835-838
76838-628
76977-902
76977-904
76977-906
77306-044
77672-588
(R)-(+)-1-(1-Naphthyl)ethylamine 98%
(R)-(+)-1-(1-Naphthyl)ethylamine
Precaución: For research use only
Formula:
C₁₂H₁₃N MW: 171.24 g/mol Boiling Pt: 289.9 °C (760 mm Hg) Flash Pt: 113 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00064114 CAS Number: 3886-70-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...