(R)-1-(4-Methoxyphenyl)ethanol 95%

Supplier: Ambeed
Warning

A148460-100MG A148460-1G
103803-930EA 39 USD
103803-930 103842-042 76823-750 77299-658
(R)-1-(4-Methoxyphenyl)ethanol 95%
(R)-1-(4-Methoxyphenyl)ethanol

Precaución: for Research Use Only

Formula: C₅H₁₃NOS
MW: 135.07 g/mol
Boiling Pt: 254.3 °C (760 mm Hg)
Flash Pt: 108 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09863667
CAS Number: 1517-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR