(R)-(+)-4-Fluoro-α-methylbenzylamine 98%

Supplier: Ambeed
Warning

Synonyms: (R)-(+)-1-(4-Fluorophenyl)ethylamine, (R)-1-(4-Fluorophenyl)ethanamine

A204035-1G A204035-25G A204035-5G A204035-10G A204035-250MG
103817-518EA 21 USD
103817-518 103848-182 77086-330 77086-332 77086-334 77139-442 77139-444
(R)-(+)-4-Fluoro-α-methylbenzylamine 98%
(R)-(+)-4-Fluoro-α-methylbenzylamine

Precaución: for Research Use Only

Formula: C₈H₁₀FN
MW: 139.17 g/mol
Boiling Pt: 185.4 °C (760 mm Hg)
Melting Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD03093090
CAS Number: 374898-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR