(R)-(+)-4-Bromo-α-methylbenzylamine 98%

Supplier: Ambeed
Danger

Synonyms: (R)-1-(4-Bromophenyl)ethylamine, (R)-(+)-1-(4-Bromophenyl)ethylamine

A150729-10G A150729-1G A150729-25G A150729-5G
103804-198EA 6 USD
103804-198 103842-336 77137-758 77299-826 77299-828 77317-864
(R)-(+)-4-Bromo-α-methylbenzylamine 98%
(R)-(+)-4-Bromo-α-methylbenzylamine

Precaución: for Research Use Only

Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Boiling Pt: 258.9 °C (760 mm Hg)
Melting Pt: …25 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00025548
CAS Number: 45791-36-4
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR