(R)-1-(3-Nitrophenyl)ethanol 97%

Supplier: Ambeed
Warning

A1003721-100MG A1003721-250MG A1003721-1G
103795-400EA 21 USD
103795-400 76646-188 76646-190 77296-652
(R)-1-(3-Nitrophenyl)ethanol 97%
(R)-1-(3-Nitrophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Flash Pt: 124 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18339582
CAS Number: 76116-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR