(R)-1-(3,4-Dimethoxyphenyl)ethanamine hydrochloride 95%

Supplier: Ambeed
Warning

A419758-1G A419758-250MG
103827-374EA 52 USD
103827-374 103838-424 76842-046 77143-140
(R)-1-(3,4-Dimethoxyphenyl)ethanamine hydrochloride 95%
(R)-1-(3,4-Dimethoxyphenyl)ethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₆ClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD24410973
CAS Number: 390815-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR