(R)-1-(2-Chlorophenyl)ethanol 97%

Supplier: Ambeed
Warning

A214122-100MG A214122-10G A214122-250MG A214122-1G A214122-5G A214122-25G
103818-370EA 20 USD
103818-370 103849-078 76999-704 76999-706 76999-708 77087-574 77087-576 77881-518
(R)-1-(2-Chlorophenyl)ethanol 97%
(R)-1-(2-Chlorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₉ClO
Boiling Pt: 231.4 °C (760 mm Hg)
Flash Pt: 94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06659510
CAS Number: 120466-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR