(R)-1-(2-Bromophenyl)ethanamine 97%

Supplier: Ambeed
Warning

A104079-100MG A104079-250MG A104079-1G
103796-648EA 91 USD
103796-648 103822-360 76809-466 76809-468 77283-192
(R)-1-(2-Bromophenyl)ethanamine 97%
(R)-1-(2-Bromophenyl)ethanamine

Precaución: for Research Use Only

Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Boiling Pt: 248.5±15.0 °C (760 mm Hg)
Flash Pt: 104.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06761910
CAS Number: 113974-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR