(1R)-1-[2-(Trifluoromethyl)phenyl]ethylamine 97%

Supplier: Ambeed
Warning

Synonyms: (R)-1-(2-(Trifluoromethyl)phenyl)ethanamine, (R)-α-Methyl-2-(trifluoromethyl)benzylamine, 2-[(1R)-1-(Aminoethyl)]benzotrifluoride

A164088-100MG A164088-250MG
103805-812EA 186 USD
103805-812 77138-266 77138-268
(1R)-1-[2-(Trifluoromethyl)phenyl]ethylamine 97%
(1R)-1-[2-(Trifluoromethyl)phenyl]ethylamine

Precaución: for Research Use Only

Formula: C₉H₁₀F₃N
MW: 189.178 g/mol
Boiling Pt: 196.3±35.0 °C (760 mm Hg)
Flash Pt: 78.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03093002
CAS Number: 127733-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR