(R)-N-Fmoc-azetidine-2-carboxylic acid 96%

Supplier: Ambeed
Warning

Synonyms: (R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)azetidine-2-carboxylic acid

A160267-250MG A160267-1G
103805-356EA 31 USD
103805-356 76812-682 77319-506
(R)-N-Fmoc-azetidine-2-carboxylic acid 96%
(R)-N-Fmoc-azetidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₉H₁₇NO₄
MW: 323.35 g/mol
Boiling Pt: 543.3 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD01320875
CAS Number: 374791-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR