(R)-(+)-2,2'-Diamino-1,1'-binaphthalene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-(+)-2,2'-Diamino-1,1'-binaphthyl, (R)-(+)-1,1'-Binaphthyl-2,2'-diamine, (R)-(+)-BINAM, (R)-[1,1'-Binaphthalene]-2,2'-diamine
A272063-100G
A272063-10G
A272063-250MG
103813-082EA
11
USD
103813-082
103852-994
77001-598
77001-600
77001-602
(R)-(+)-2,2'-Diamino-1,1'-binaphthalene 98%
(R)-(+)-2,2'-Diamino-1,1'-binaphthalene
Precaución: for Research Use Only
Formula:
C₂₀H₁₆N₂ MW: 284.362 g/mol Boiling Pt: 480.8 °C (760 mm Hg) Melting Pt: 242…244 °C Flash Pt: 293.854 °C Storage Temperature: Ambient |
MDL Number:
MFCD00145204 CAS Number: 18741-85-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...