(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 98%

Supplier: Ambeed
Warning

Synonyms: (R)-(-)-1,2-O-Isopropylidenglycerol, (R)-(-)-2,2-Dimethyl-1,3-dioxolane-4-methanol, (R)-(-)-2,3-O-Isopropylideneglycerol, (R)-(-)-1,2-O-Isopropylideneglycerol, (R)-(2,2-Dimethyl-1,3-dioxolan-4-yl)methanol

A184850-100G A184850-10G A184850-500G A184850-25G A184850-5G A184850-1G
103815-798EA 9 USD
103815-798 103846-364 76998-808 76998-810 76998-812 77083-990 77083-992 77301-928
(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 98%
(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol

Precaución: for Research Use Only

Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 188.5 °C (760 mm Hg)
Melting Pt: …13.7 °C
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00003213
CAS Number: 14347-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR