(R)-(1,4-Dioxan-2-yl)methanamine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A288467-1G
A288467-5G
A288467-100MG
A288467-250MG
A288467-25G
103813-798EA
94
USD
103813-798
103834-940
76837-500
76837-502
77153-948
77153-950
77153-952
(R)-(1,4-Dioxan-2-yl)methanamine hydrochloride 97%
(R)-(1,4-Dioxan-2-yl)methanamine hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₂ClNO₂ MW: 153.6 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD27918519 CAS Number: 1523541-84-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...