(R)-1,2-Diaminopropane dihydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-(+)-1,2-Diaminopropane dihydrochloride, (R)-1,2-Propanediamine Dihydrochloride, (R)-Propylenediamine Dihydrochloride
A153042-100G
A153042-10G
A153042-250MG
A153042-25G
A153042-5G
A153042-1G
103804-460EA
15
USD
103804-460
103842-632
77137-862
77137-864
77137-866
77299-994
77299-996
77318-272
(R)-1,2-Diaminopropane dihydrochloride 97%
(R)-1,2-Diaminopropane dihydrochloride
Precaución: for Research Use Only
Formula:
C₃H₁₀N₂ Melting Pt: 241…244 °C Flash Pt: 84.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00239481 CAS Number: 19777-67-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...