(R)-Ibuprofen 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-2-(4-Isobutylphenyl)propanoic acid, (R)-(-)-Ibuprofen
A275175-1G
A275175-100MG
A275175-250MG
A275175-5G
103813-238EA
102
USD
103813-238
77093-006
77141-032
77571-694
77746-488
(R)-Ibuprofen 96%
(R)-Ibuprofen
Precaución: for Research Use Only
Formula:
C₁₃H₁₈O₂ MW: 206.3 g/mol Boiling Pt: 319.6 °C (760 mm Hg) Melting Pt: 41…42 °C Flash Pt: 216.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00069290 CAS Number: 51146-57-7 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...