(R)-(-)-5-Oxotetrahydrofuran-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A113469-100G A113469-10G A113469-250MG A113469-25G A113469-1G A113469-5G
103798-236EA 24 USD
103798-236 103824-078 77136-130 77136-132 77136-134 77136-136 77297-246 77310-894
(R)-(-)-5-Oxotetrahydrofuran-2-carboxylic acid 95%
(R)-(-)-5-Oxotetrahydrofuran-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₆O₄
Boiling Pt: 384.2 °C (760 mm Hg)
Melting Pt: 69…73 °C
Flash Pt: 184.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00011567
CAS Number: 53558-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR