(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone 98%
Supplier: Ambeed
|
Warning
|
A555150-100G
A555150-10G
A555150-250MG
A555150-1G
A555150-25G
A555150-5G
A555150-500G
103828-708EA
6
USD
103828-708
103839-844
76819-088
76819-090
76819-092
76843-934
76843-936
76843-938
77613-746
(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone 98%
(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone
Precaución: for Research Use Only
Formula:
C₅H₉NO₂ MW: 115.13 g/mol Boiling Pt: 346 °C (760 mm Hg) Melting Pt: 83…85 °C Flash Pt: 163 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077791 CAS Number: 66673-40-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...