(R)-(-)-4-Benzyl-3-propionyl-2-oxazolidinone 97%

Supplier: Ambeed
Warning

A516293-1G A516293-25G A516293-10G A516293-250MG
103828-452EA 19 USD
103828-452 103839-558 76843-544 76843-546 77004-804 77004-806
(R)-(-)-4-Benzyl-3-propionyl-2-oxazolidinone 97%
(R)-(-)-4-Benzyl-3-propionyl-2-oxazolidinone

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₃
MW: 233.2631 g/mol
Boiling Pt: 396.6 °C (760 mm Hg)
Melting Pt: 45 °C
Flash Pt: 193.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00269687
CAS Number: 131685-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR