(R)-(-)-3-Fluoropyrrolidine hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-(-)-3-Fluoropyrrolidine Hydrochloride
A173142-100G
A173142-100MG
A173142-10G
A173142-250MG
A173142-1G
A173142-25G
A173142-5G
103806-796EA
10
USD
103806-796
103845-090
77011-664
77011-666
77011-668
77011-670
77082-322
77082-324
77082-326
(R)-(-)-3-Fluoropyrrolidine hydrochloride 98%
(R)-(-)-3-Fluoropyrrolidine hydrochloride
Precaución: for Research Use Only
Formula:
C₄H₉ClFN MW: 125.57 g/mol Melting Pt: 179…186 °C Storage Temperature: Ambient |
MDL Number:
MFCD04038717 CAS Number: 136725-55-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...