(R)-(-)-1-Cbz-3-Pyrrolidinol 98%

Supplier: Ambeed
Danger

Synonyms: (R)-1-Cbz-3-Pyrrolidinol, (R)-1-Carbobenzoxy-3-pyrrolidinol, Benzyl (R)-3-hydroxypyrrolidine-1-carboxylate, (R)-1-Benzyloxycarbonyl-3-hydroxypyrrolidine, (R)-1-Cbz-3-pyrrolidinol, (R)-1-Benzyloxycarbonyl-3-pyrrolidinol, (R)-1-Carbobenzoxy-3-hydroxypyrrolidine

A172077-1G A172077-10G A172077-25G A172077-5G
103806-692EA 9 USD
103806-692 103844-978 77082-154 77138-440 77301-184 77301-186
(R)-(-)-1-Cbz-3-Pyrrolidinol 98%
(R)-(-)-1-Cbz-3-Pyrrolidinol

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₃
MW: 221.2524 g/mol
Melting Pt: 74…79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368258
CAS Number: 100858-33-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR