Bis[2-(diphenylphosphino)phenyl]ether 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,2'-Bis(diphenylphosphino)diphenyl Ether, DPEphos, Bis(2-diphenylphosphinophenyl)ether, (Oxydi-2,1-phenylene)bis(diphenylphosphine), (Oxybis(2,1-phenylene))bis(diphenylphosphine)
A152563-100G
A152563-10G
A152563-500G
A152563-25G
A152563-5G
103804-404EA
5
USD
103804-404
103842-576
77137-830
77137-832
77137-834
77299-960
77318-196
Bis[2-(diphenylphosphino)phenyl]ether 97%
Bis[2-(diphenylphosphino)phenyl]ether
Precaución: for Research Use Only
Formula:
C₃₆H₂₈OP₂ Boiling Pt: 608.2 °C (760 mm Hg) Melting Pt: 184…187 °C Flash Pt: 404.905 °C Storage Temperature: Ambient |
MDL Number:
MFCD00233863 CAS Number: 166330-10-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...