(Oxybis(2,1-phenylene))bis(dicyclohexylphosphine) 98%

Supplier: Ambeed
Warning

A485705-100G A485705-1G A485705-25G A485705-5G
103828-172EA 8 USD
103828-172 103839-246 76830-016 76843-184 76843-186 76843-188
(Oxybis(2,1-phenylene))bis(dicyclohexylphosphine) 98%
(Oxybis(2,1-phenylene))bis(dicyclohexylphosphine)

Precaución: for Research Use Only

Formula: C₃₆H₅₂OP₂
MW: 562.75 g/mol
Boiling Pt: 652.6±40.0 °C (760 mm Hg)
Melting Pt: 140…146 °C
Flash Pt: 439.6 °C
Storage Temperature: Ambient
MDL Number: MFCD03058784
CAS Number: 434336-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR