(Methoxymethyl)triphenylphosphonium chloride 98%

Supplier: Ambeed
Warning

A101931-100G A101931-1G A101931-25G A101931-500G A101931-1000G
103807-830EA 55 USD
103807-830 76970-192 76970-194 76970-196 76970-198 77308-792
(Methoxymethyl)triphenylphosphonium chloride 98%
(Methoxymethyl)triphenylphosphonium chloride

Precaución: for Research Use Only

Formula: C₂₀H₂₀ClOP
MW: 342.8 g/mol
Melting Pt: 185…195 °C
Flash Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD00011800
CAS Number: 4009-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR