Benzyl methyl ether 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
(Methoxymethyl)benzene
A372710-25G
A372710-5G
103837-532EA
7
USD
103837-532
76840-794
76840-796
Benzyl methyl ether 98%
Benzyl methyl ether
Precaución: for Research Use Only
Formula:
C₈H₁₀O MW: 122.17 g/mol Boiling Pt: 174 °C (760 mm Hg) Melting Pt: 52.6 °C Flash Pt: 36.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00025901 CAS Number: 538-86-3 UN: 3271 ADR: 3,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...