(Iodomethyl)trimethylsilane 97%

Supplier: Ambeed
Danger

A493482-25G A493482-5G
103839-310EA 32 USD
103839-310 76843-272 76843-274
(Iodomethyl)trimethylsilane 97%
(Iodomethyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₄H₁₁ISi
MW: 214.12 g/mol
Boiling Pt: 140 °C (760 mm Hg)
Flash Pt: 31.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001077
CAS Number: 4206-67-1
UN: 2924
ADR: 3,8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR