(2R,2'R)-Dimethyl-3,3'-disulfanediylbis(2-aminopropanoate) dihydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(H-Cystine-OMe)₂·2HCl, L-Cystine dimethyl ester dihydrochloride, (H-Cys-OMe)₂·2 HCl, (H-Cys-OMe)2.2HCl
A116644-25G
A116644-500G
A116644-1G
A116644-5G
A116644-100G
A116644-10G
103798-776EA
5
USD
103798-776
103824-640
76995-548
76995-550
77297-482
77297-484
77670-630
77670-632
(2R,2'R)-Dimethyl-3,3'-disulfanediylbis(2-aminopropanoate) dihydrochloride 97%
(2R,2'R)-Dimethyl-3,3'-disulfanediylbis(2-aminopropanoate) dihydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₈Cl₂N₂O₄S₂ MW: 341.28 g/mol Boiling Pt: 375.5 °C (760 mm Hg) Melting Pt: 182…183 °C Flash Pt: 180.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012490 CAS Number: 32854-09-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...