(E)-Methyl 4-(dimethylamino)but-2-enoate 95%
Supplier: Ambeed
|
Warning
|
A687783-100MG
A687783-1G
103840-562EA
766
USD
103840-562
76651-066
76844-780
(E)-Methyl 4-(dimethylamino)but-2-enoate 95%
(E)-Methyl 4-(dimethylamino)but-2-enoate
Precaución: for Research Use Only
Formula:
C₇H₁₃NO₂ Flash Pt: 63 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD18207077 CAS Number: 212776-19-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...