Methyl (3E)-4-(4-bromophenyl)-2-oxobut-3-enoate 95%

Supplier: Ambeed
Warning

Synonyms: (E)-Methyl 4-(4-bromophenyl)-2-oxobut-3-enoate

A139136-250MG A139136-5G A139136-1G
103802-698EA 90 USD
103802-698 76811-672 76811-674 77315-778
Methyl (3E)-4-(4-bromophenyl)-2-oxobut-3-enoate 95%
Methyl (3E)-4-(4-bromophenyl)-2-oxobut-3-enoate

Precaución: for Research Use Only

Formula: C₁₁H₉BrO₃
MW: 269.091 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18642374
CAS Number: 608128-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR