(E)-Methyl 3-(3,5-difluoro-4-formylphenyl)acrylate 97%

Supplier: Ambeed
Warning

A331109-1G A331109-5G A331109-250MG
103825-570EA 41 USD
103825-570 103836-534 76839-516 76839-518 77239-766
(E)-Methyl 3-(3,5-difluoro-4-formylphenyl)acrylate 97%
(E)-Methyl 3-(3,5-difluoro-4-formylphenyl)acrylate

Precaución: for Research Use Only

Formula: C₁₁H₈F₂O₃
MW: 226.18 g/mol
Boiling Pt: 320.9±42.0 °C (760 mm Hg)
Flash Pt: 143.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD29905622
CAS Number: 1639042-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR