Ethyl-trans-4-oxo-2-butenoate 96%
Supplier: Ambeed
|
Danger
|
Synonyms:
(E)-Ethyl 4-oxobut-2-enoate
A134324-10G
A134324-1G
A134324-25G
A134324-5G
103802-002EA
24
USD
103802-002
76797-990
77314-736
77314-738
77314-740
Ethyl-trans-4-oxo-2-butenoate 96%
Ethyl-trans-4-oxo-2-butenoate
Precaución: for Research Use Only
Formula:
C₆H₈O₃ MW: 128.13 g/mol Boiling Pt: 199.3 °C (760 mm Hg) Melting Pt: 9 °C Flash Pt: 68 °C Storage Temperature: Freezer |
MDL Number:
MFCD00236170 CAS Number: 2960-66-9 UN: 2922 ADR: 8,6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...