Ethyl-4-bromocrotonate 84%

Supplier: Ambeed
Danger

Synonyms: (E)-Ethyl 4-bromobut-2-enoate, Ethyl trans-4-bromo-2-butenoate

A163098-100G A163098-10G A163098-500G A163098-5G
103843-946EA 10 USD
103843-946 76812-850 76812-852 76812-854 77300-634
Ethyl-4-bromocrotonate 84%
Ethyl-4-bromocrotonate

Precaución: for Research Use Only

Formula: C₆H₉BrO₂
Boiling Pt: 213.9 °C (760 mm Hg)
Flash Pt: 97 °C
Storage Temperature: Freezer
MDL Number: MFCD00000247
CAS Number: 37746-78-4
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR