[(E)-2-(Ethoxycarbonyl)vinyl]boronic acid, pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: [(E)-2-(Ethoxycarbonyl)ethenyl]boronic acid, pinacol ester, (E)-Ethyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)acrylate, Ethyl (2E)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)prop-2-enoate, Ethyl (E)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)acrylate

A239843-100MG A239843-10G A239843-250MG A239843-1G A239843-25G A239843-5G
103811-312EA 11 USD
103811-312 103851-110 76815-376 76815-378 76815-380 77090-252 77090-254 77090-256
[(E)-2-(Ethoxycarbonyl)vinyl]boronic acid, pinacol ester 97%
[(E)-2-(Ethoxycarbonyl)vinyl]boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₉BO₄
Storage Temperature: Refrigerator
MDL Number: MFCD09027313
CAS Number: 1009307-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR