(E)-Ethyl-2-cyano-3-ethoxyacrylate 99%

Supplier: Ambeed
Danger

A172758-100G A172758-10G A172758-25G A172758-1G A172758-5G
103806-748EA 5 USD
103806-748 103845-040 76974-092 76974-094 76974-096 77301-216 77301-218
(E)-Ethyl-2-cyano-3-ethoxyacrylate 99%
(E)-Ethyl-2-cyano-3-ethoxyacrylate

Precaución: for Research Use Only

Formula: C₈H₁₁NO₃
Melting Pt: 52 °C
Storage Temperature: Ambient
MDL Number: MFCD00009136
CAS Number: 42466-67-1
UN: 3335
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR