trans-Cinnamonitrile 97%

Supplier: Ambeed
Warning

Synonyms: (E)-3-Phenylacrylonitrile, Cinnamonitrile

A550815-25G A550815-5G
103839-820EA 5 USD
103839-820 76843-890 77307-794
trans-Cinnamonitrile 97%
trans-Cinnamonitrile

Precaución: for Research Use Only

Formula: C₉H₇N
MW: 129.16 g/mol
Boiling Pt: 263.8 °C (760 mm Hg)
Melting Pt: 18…20 °C
Flash Pt: 105.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00001930
CAS Number: 1885-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR