trans-Chalcone 97%

Supplier: Ambeed
Warning

Synonyms: trans-Benzylidenacetophenone, (E)-Chalcone, trans-Benzalacetophenone, (E)-1,3-Diphenylprop-2-en-1-one

A131628-100G A131628-10G A131628-500G A131628-25G
103808-430EA 167 USD
103808-430 76870-768 76870-770 76870-772 77314-118
trans-Chalcone 97%
trans-Chalcone

Precaución: for Research Use Only

Formula: C₁₅H₁₂O
MW: 208.26 g/mol
Boiling Pt: 346.6 °C (760 mm Hg)
Melting Pt: 55…59 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003082
CAS Number: 614-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR