Fumaric acid monoethyl ester 98%

Supplier: Ambeed
Warning

Synonyms: (E)-4-Ethoxy-4-oxobut-2-enoic acid, Monoethyl fumarate, Ethyl hydrogen fumarate

A145377-100G A145377-500G A145377-25G
103841-614EA 7 USD
103841-614 76767-972 76767-974 77299-450
Fumaric acid monoethyl ester 98%
Fumaric acid monoethyl ester

Precaución: for Research Use Only

Formula: C₆H₈O₄
MW: 144.13 g/mol
Boiling Pt: 261.6 °C (760 mm Hg)
Melting Pt: 66…68 °C
Flash Pt: 109.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00002699
CAS Number: 2459-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR