4-Bromocrotonic acid 95%
Supplier: Ambeed
|
Danger
|
A698521-1G
A698521-25G
A698521-5G
A698521-100G
A698521-10G
A698521-250MG
A698521-500G
103840-618EA
16
USD
103840-618
76844-844
76844-846
76844-848
77293-280
77293-282
77293-284
77293-286
4-Bromocrotonic acid 95%
4-Bromocrotonic acid
Precaución: for Research Use Only
Formula:
C₄H₅BrO₂ Boiling Pt: 287.6 °C (760 mm Hg) Melting Pt: 74 °C Flash Pt: 127.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00082701 CAS Number: 13991-36-1 UN: 3261 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...