trans-4-(Trifluoromethyl)cinnamic acid 98%

Supplier: Ambeed
Warning

A176304-100G A176304-10G A176304-1G A176304-25G A176304-5G A176304-500G
103845-454EA 7 USD
103845-454 76769-418 76769-420 76769-422 77082-804 77301-446 77877-256
trans-4-(Trifluoromethyl)cinnamic acid 98%
trans-4-(Trifluoromethyl)cinnamic acid

Precaución: for Research Use Only

Formula: C₁₀H₇F₃O₂
Boiling Pt: 279.2 °C (760 mm Hg)
Melting Pt: 230…234 °C
Flash Pt: 122.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00002696
CAS Number: 16642-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR