(E)-4-(tert-Butoxy)-4-oxobut-2-enoic acid 95%

Supplier: Ambeed
Warning

A419369-1G A419369-5G A419369-10G A419369-250MG A419369-25G
103827-368EA 41 USD
103827-368 103838-414 76842-028 76842-030 77168-512 77168-514 77168-516
(E)-4-(tert-Butoxy)-4-oxobut-2-enoic acid 95%
(E)-4-(tert-Butoxy)-4-oxobut-2-enoic acid

Precaución: for Research Use Only

Formula: C₈H₁₂O₄
MW: 172.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08669525
CAS Number: 135355-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR