Piceatannol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(E)-4-(3,5-Dihydroxystyryl)benzene-1,2-diol
A162917-100MG
A162917-10G
A162917-10MG
A162917-250MG
A162917-25G
A162917-25MG
A162917-50MG
A162917-1G
A162917-5G
103805-668EA
61
USD
103805-668
103843-926
77028-676
77028-678
77028-680
77028-682
77028-684
77028-686
77028-688
77300-620
77300-622
Piceatannol 98%
Piceatannol
Precaución: for Research Use Only
Formula:
C₁₄H₁₂O₄ Melting Pt: 223…227 °C Flash Pt: 252.2 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00221715 CAS Number: 10083-24-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...