(E)-4-(2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)vinyl)benzonitrile 97%

Supplier: Ambeed
Warning

A128350-100MG A128350-250MG
103801-112EA 216 USD
103801-112 76971-958 77566-494
(E)-4-(2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)vinyl)benzonitrile 97%
(E)-4-(2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)vinyl)benzonitrile

Precaución: For research use only.

Formula: C₁₅H₁₈BNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD19703873
CAS Number: 172512-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR