trans-4-Bromocinnamaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: (E)-3-(4-Bromophenyl)acrylaldehyde

A899672-1G A899672-5G A899672-25G
103841-346EA 24 USD
103841-346 76845-612 76845-614 77294-168
trans-4-Bromocinnamaldehyde 97%
trans-4-Bromocinnamaldehyde

Precaución: for Research Use Only

Formula: C₉H₇BrO
MW: 211.06 g/mol
Boiling Pt: 310.5 °C (760 mm Hg)
Melting Pt: 78…82 °C
Flash Pt: 114.5 °C
Storage Temperature: Freezer
MDL Number: MFCD03032509
CAS Number: 49678-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR