(E)-3-(4-(Benzyloxy)phenyl)acrylic acid 99%

Supplier: Ambeed
Warning

A326181-1G A326181-10G A326181-5G
103825-420EA 12 USD
103825-420 103836-368 76839-316 77114-812 77306-332
(E)-3-(4-(Benzyloxy)phenyl)acrylic acid 99%
(E)-3-(4-(Benzyloxy)phenyl)acrylic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₄O₃
Boiling Pt: 448.9 °C (760 mm Hg)
Melting Pt: 206…208 °C
Flash Pt: 169.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00021778
CAS Number: 227105-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR